siRNA Details
| Gene Symbol: C4bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007576.3 | NP_031602.3 | 4, 5 | 637 | ||
| GenBank mRNA | AB094485.1 | - | 618 | |||
| AB094486.1 | - | 4, 5 | 553 | |||
| AK136735.1 | - | 4, 5 | 551 | |||
| AK149468.1 | - | 4, 5 | 644 | |||
| AK149477.1 | - | 4, 5 | 554 | |||
| AK162345.1 | - | 4, 5 | 551 | |||
| BC012257.1 | - | 4, 5 | 609 | 4163203 | ||
| M17122.1 | - | 4, 5 | 651 | |||

C4bp
Genomic Map