siRNA Details
| Gene Symbol: Bmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007555.3 | NP_031581.2 | 1 | 947 | ||
| GenBank mRNA | AK033362.1 | - | 1 | 950 | ||
| AK160412.1 | - | 55 | ||||
| BC100751.1 | - | 1 | 218 | |||
| BC100752.1 | - | 1 | 218 | |||
| BC100753.1 | - | 1 | 218 | |||
| BC100754.1 | - | 1 | 218 | |||
| BC141283.1 | - | 1 | 750 | 9055895 | ||
| L41145.1 | - | 1 | 219 | |||

Bmp5
Genomic Map