siRNA Details
| Gene Symbol: Bmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007556.2 | NP_031582.1 | 6 | 1519 | ||
| GenBank mRNA | AK014797.1 | - | 57 | |||
| AK041210.1 | - | 1877 | ||||
| AK135770.1 | - | 6 | 1152 | |||
| BC138593.1 | - | 6 | 1439 | 8861613 | ||
| BC138595.1 | - | 6 | 1439 | 8861615 | ||
| J04566.1 | - | 6 | 1101 | |||
| X80992.1 | - | 6 | 1518 | |||

Bmp6
Genomic Map