siRNA Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243645.1 | NP_001230574.1 | 2 | 276 | ||
| NM_021914.7 | NP_068733.1 | 2 | 299 | |||
| NM_138638.4 | NP_619579.1 | 2 | 441 | |||
| NR_028130.1 | 2 | 441 | ||||
| XM_011536363.2 | XP_011534665.1 | 2 | 237 | |||
| GenBank mRNA | AF087867.1 | - | 2 | 194 | ||
| AF134802.1 | - | 2 | 310 | |||
| AF134803.1 | - | 2 | 204 | |||
| AK314157.1 | - | 2 | 259 | |||
| AL599958.1 | - | 2 | 276 | |||
| BC011444.1 | - | 2 | 225 | 4245104 | ||
| BC022364.1 | - | 2 | 308 | 4730117 | ||
| BC022876.1 | - | 2 | 312 | 4109108 | ||
| CB992605.1 | - | 2 | 284 | |||
| DA298739.1 | - | 2 | 264 | |||
| DA897112.1 | - | 2 | 276 | |||

CFL2
Genomic Map