siRNA Details
| Gene Symbol: Arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB006103.2 | - | 2, 3 | 276 | ||
| BC052033.1 | - | 1, 2 | 270 | 5707995 | ||
| CJ045476.1 | - | 1, 2 | 409 | |||
| CJ049350.1 | - | 2, 3 | 290 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arx| Gene Symbol: Arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB006103.2 | - | 2, 3 | 276 | ||
| BC052033.1 | - | 1, 2 | 270 | 5707995 | ||
| CJ045476.1 | - | 1, 2 | 409 | |||
| CJ049350.1 | - | 2, 3 | 290 | |||