siRNA Details
| Gene Symbol: Astn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205204.1 | NP_001192133.1 | 23 | 5156 | ||
| NM_007495.4 | NP_031521.2 | 23 | 5132 | |||
| GenBank mRNA | AB093222.2 | - | 23 | 4700 | ||
| AK147450.1 | - | 23 | 4902 | |||
| BC054545.1 | - | 20 | 3906 | |||
| BC094666.1 | - | 23 | 5018 | 30545499 | ||
| G49230.1 | - | 23 | 4738 | |||
| U48797.1 | - | 23 | 4738 | |||

Astn1
Genomic Map