siRNA Details
| Gene Symbol: Bcap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164090.1 | NP_001157562.1 | 8 | 1445 | ||
| NM_007530.3 | NP_031556.1 | 7 | 1608 | |||
| GenBank mRNA | AK090068.1 | - | 8 | 1422 | ||
| AK144754.1 | - | 8 | 1419 | |||
| AK205917.1 | - | 1 | 48 | |||
| AK216433.1 | - | 1 | 42 | |||
| BC003303.1 | - | 7 | 1512 | 3584161 | ||
| BC021661.1 | - | 7 | 1619 | 2646883 | ||
| BC040751.1 | - | 2 | 1798 | |||
| X78684.1 | - | 7 | 1419 | |||

Bcap29
Genomic Map