siRNA Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007542.4 | NP_031568.2 | 8 | 2150 | ||
| GenBank mRNA | AK155973.1 | - | 8 | 2141 | ||
| BC005452.1 | - | 8 | 2134 | 3594469 | ||
| BC019502.1 | - | 8 | 2143 | 4237268 | ||
| BC052857.1 | - | 8 | 2125 | 30061850 | ||
| L20276.1 | - | 8 | 2140 | |||

Bgn
Genomic Map