siRNA Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007644.3 | NP_031670.1 | 2 | 452 | ||
| GenBank mRNA | AB008553.1 | - | 2 | 357 | ||
| AK011123.1 | - | 2 | 382 | |||
| AK083038.1 | - | 2 | 453 | |||
| AK144235.1 | - | 2 | 484 | |||
| AK149102.1 | - | 398 | ||||
| BC029073.1 | - | 2 | 360 | 3962669 | ||

Scarb2
Genomic Map