siRNA Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007651.3 | NP_031677.1 | 3 | 435 | ||
| GenBank mRNA | AA177409.1 | - | 3 | 323 | ||
| AK156510.1 | - | 3 | 342 | |||
| AK160326.1 | - | 3 | 333 | |||
| BC021310.1 | - | 3 | 349 | 5010267 | ||
| BC052905.1 | - | 3 | 300 | 30078163 | ||
| X97227.1 | - | 2 | 236 | |||
| Z16071.1 | - | 2 | 221 | |||

Cd53
Genomic Map