siRNA Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110320.1 | NP_001103790.1 | 1, 2 | 188 | ||
| NM_001110321.1 | NP_001103791.1 | 1, 2 | 188 | |||
| NM_001110322.1 | NP_001103792.1 | 1, 2 | 188 | |||
| NM_007654.2 | NP_031680.2 | 1, 2 | 188 | |||
| GenBank mRNA | AF543213.1 | - | 1, 2 | 89 | ||
| AK134132.1 | - | 1, 2 | 191 | |||
| AK144191.1 | - | 1, 2 | 191 | |||
| AK151094.1 | - | 190 | ||||
| BC003824.1 | - | 1, 2 | 146 | 3599554 | ||
| BY743956.1 | - | 1, 2 | 188 | |||
| J04170.1 | - | 1, 2 | 125 | |||
| S40777.1 | - | 1, 2 | 121 | |||

Cd72
Genomic Map