siRNA Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007658.3 | NP_031684.3 | 5 | 778 | ||
| GenBank mRNA | AK077609.1 | - | 5 | 395 | ||
| AK077991.1 | - | 5 | 781 | |||
| AK142334.1 | - | 4 | 1053 | |||
| BC046296.1 | - | 5 | 397 | 6486343 | ||
| U27323.1 | - | 5 | 364 | |||

Cdc25a
Genomic Map