siRNA Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007673.3 | NP_031699.2 | 2 | 881 | ||
| GenBank mRNA | AK049566.1 | - | 2 | 882 | ||
| AK049574.1 | - | 2 | 753 | |||
| BC103516.1 | - | 2 | 759 | 40039012 | ||
| BC103517.1 | - | 2 | 762 | 40039015 | ||
| BC103519.1 | - | 2 | 762 | 40039019 | ||
| BC103520.1 | - | 2 | 762 | 40039017 | ||
| S74520.1 | - | 2 | 876 | |||

Cdx2
Genomic Map