siRNA Details
| Gene Symbol: Cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007688.2 | NP_031714.1 | 2 | 220 | ||
| GenBank mRNA | AK004659.1 | - | 2 | 212 | ||
| AK147174.1 | - | 2 | 134 | |||
| BC007138.1 | - | 2 | 163 | 3590849 | ||
| L29468.1 | - | 2 | 220 | |||

Cfl2
Genomic Map