siRNA Details
| Gene Symbol: Chgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007694.4 | NP_031720.1 | 4 | 665 | ||
| GenBank mRNA | AK019893.1 | - | 4 | 275 | ||
| AK077487.1 | - | 4 | 231 | |||
| AK077489.1 | - | 4 | 273 | |||
| AK133629.1 | - | 4 | 273 | |||
| AK133739.1 | - | 4 | 273 | |||
| AK133786.1 | - | 4 | 275 | |||
| AK158857.1 | - | 4 | 666 | |||
| BC014736.1 | - | 4 | 266 | 4511248 | ||
| X51429.1 | - | 4 | 261 | |||
| X53028.1 | - | 4 | 271 | |||

Chgb
Genomic Map