siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301250.1 | NP_001288179.1 | 8 | 1248 | ||
| NM_007602.4 | NP_031628.1 | 8 | 1412 | |||
| GenBank mRNA | AK164293.1 | - | 8 | 1322 | ||
| AK170034.1 | - | 8 | 1349 | |||
| BC014767.1 | - | 8 | 1240 | 4458488 | ||
| U85020.1 | - | 8 | 1331 | |||
| Y10656.1 | - | 8 | 1344 | |||

Capn5
Genomic Map