siRNA Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042072.1 | - | 2 | 214 | ||
| AK052396.1 | - | 1 | 2566 | |||
| AK133829.1 | - | 2 | 223 | |||
| AK197928.1 | - | 1 | 142 | |||
| AK199368.1 | - | 2 | 142 | |||
| AK210521.1 | - | 2 | 142 | |||
| BC034262.1 | - | 2 | 280 | 4460089 | ||
| BY074786.1 | - | 2 | 336 | |||
| D28492.1 | - | 2 | 203 | |||
| Y13085.1 | - | 2 | 282 | |||

Casp2
Genomic Map