siRNA Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007611.2 | NP_031637.1 | 7 | 1290 | ||
| GenBank mRNA | AK079250.1 | - | 2 | 577 | ||
| AK081718.1 | - | 1 | 335 | |||
| AK154978.1 | - | 7 | 1171 | |||
| BC005428.1 | - | 7 | 1147 | 3600155 | ||
| Y13088.1 | - | 7 | 1290 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U67321.1 | - | 1684 | Chr. - - [-] on Build | ||

Casp7
Genomic Map