siRNA Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007630.2 | NP_031656.2 | 7, 8 | 1089 | ||
| GenBank mRNA | AK013312.1 | - | 7, 8 | 1058 | ||
| AK076122.1 | - | 7, 8 | 1083 | |||
| BC008247.1 | - | 7, 8 | 1005 | 3156950 | ||
| CT010286.1 | - | 7, 8 | 975 | |||
| X66032.1 | - | 7, 8 | 1076 | |||

Ccnb2
Genomic Map