siRNA Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007633.2 | NP_031659.2 | 6 | 553 | ||
| GenBank mRNA | AK089950.1 | - | 6 | 485 | ||
| AK163046.1 | - | 6 | 542 | |||
| AK165658.1 | - | 6 | 543 | |||
| BC003499.1 | - | 2 | 105 | |||
| BC062152.1 | - | 6 | 580 | 6510163 | ||
| BC084588.1 | - | 6 | 597 | 6506897 | ||
| BC106191.1 | - | 6 | 553 | 6336006 | ||
| BC138660.1 | - | 5 | 386 | 8861680 | ||
| BC138662.1 | - | 5 | 386 | 8861683 | ||
| X75888.1 | - | 5 | 393 | |||

Ccne1
Genomic Map