siRNA Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007644.3 | NP_031670.1 | 3 | 599 | ||
| GenBank mRNA | AB008553.1 | - | 3 | 504 | ||
| AK011123.1 | - | 3 | 529 | |||
| AK083038.1 | - | 3 | 600 | |||
| AK144235.1 | - | 3 | 631 | |||
| AK149102.1 | - | 545 | ||||
| BC029073.1 | - | 3 | 507 | 3962669 | ||

Scarb2
Genomic Map