siRNA Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290802.1 | NP_001277731.1 | 7 | 802 | ||
| NM_001290804.1 | NP_001277733.1 | 6 | 727 | |||
| NM_007645.4 | NP_031671.1 | 6 | 751 | |||
| GenBank mRNA | AK154250.1 | - | 6 | 791 | ||
| AK154466.1 | - | 7 | 772 | |||
| AK155663.1 | - | 6 | 728 | |||
| AK155966.1 | - | 6 | 716 | |||
| BC019402.1 | - | 6 | 788 | 4013070 | ||
| CT010217.1 | - | 6 | 640 | |||

Cd37
Genomic Map