siRNA Details
| Gene Symbol: Cd3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007648.4 | NP_031674.1 | 8 | 1423 | ||
| GenBank mRNA | J02990.1 | - | 8 | 1424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd3e| Gene Symbol: Cd3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007648.4 | NP_031674.1 | 8 | 1423 | ||
| GenBank mRNA | J02990.1 | - | 8 | 1424 | ||