siRNA Details
| Gene Symbol: GPRC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018654.1 | NP_061124.1 | 2 | 929 | ||
| XM_005253421.3 | XP_005253478.1 | 4 | 3369 | |||
| XM_017019582.1 | XP_016875071.1 | 3 | 1462 | |||
| GenBank mRNA | AF209923.1 | - | 2 | 929 | ||
| BC069341.1 | - | 2 | 929 | 7262221 | ||
| BC107077.1 | - | 2 | 929 | 40027061 | ||
| BC107078.1 | - | 2 | 929 | 40027066 | ||
More Information
Set Membership: |
|

GPRC5D
Genomic Map