siRNA Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007661.3 | NP_031687.2 | 10 | 1126 | ||
| GenBank mRNA | AK077668.1 | - | 10 | 1127 | ||
| AK147133.1 | - | 10 | 1104 | |||
| BC052920.1 | - | 10 | 1204 | 6489832 | ||
| L37092.1 | - | 9 | 1698 | |||

Cdk11b
Genomic Map