siRNA Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007673.3 | NP_031699.2 | 2, 3 | 959 | ||
| GenBank mRNA | AK049566.1 | - | 2, 3 | 960 | ||
| AK049574.1 | - | 2, 3 | 831 | |||
| BC103516.1 | - | 2, 3 | 837 | 40039012 | ||
| BC103517.1 | - | 2, 3 | 840 | 40039015 | ||
| BC103519.1 | - | 2, 3 | 840 | 40039019 | ||
| BC103520.1 | - | 2, 3 | 840 | 40039017 | ||
| S74520.1 | - | 2, 3 | 954 | |||

Cdx2
Genomic Map