siRNA Details
| Gene Symbol: Cetn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007684.3 | NP_031710.1 | 5 | 528 | ||
| GenBank mRNA | AK003473.1 | - | 5 | 544 | ||
| AK004532.1 | - | 5 | 540 | |||
| AK006289.1 | - | 5 | 543 | |||
| AK006891.1 | - | 5 | 528 | |||
| AK011821.1 | - | 5 | 521 | |||
| AK017505.1 | - | 5 | 568 | |||
| BC002162.1 | - | 5 | 562 | 3485864 | ||
| Y12474.1 | - | 5 | 479 | |||

Cetn3
Genomic Map