siRNA Details
Gene Symbol: Chrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112697.1 | NP_001106167.1 | 3 | 3739 | ||
NM_007698.3 | NP_031724.2 | 2 | 3647 | |||
GenBank mRNA | AK081248.1 | - | 2 | 3304 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
