siRNA Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF033566.1 | - | 2 | 181 | ||
| AY653308.1 | - | 1 | 103 | |||
| BC012675.1 | - | 2 | 203 | 3982920 | ||
| CT010293.1 | - | 1 | 103 | |||
| U94846.1 | - | 2 | 200 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clk4| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF033566.1 | - | 2 | 181 | ||
| AY653308.1 | - | 1 | 103 | |||
| BC012675.1 | - | 2 | 203 | 3982920 | ||
| CT010293.1 | - | 1 | 103 | |||
| U94846.1 | - | 2 | 200 | |||