siRNA Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007788.3 | NP_031814.2 | 13 | 1711 | ||
| GenBank mRNA | AK146032.1 | - | 13 | 1670 | ||
| BC026149.1 | - | 13 | 1616 | 4188101 | ||
| BC060742.1 | - | 13 | 1664 | 4945773 | ||
| BC089343.1 | - | 13 | 1588 | 30360017 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006235221, XM_006235220, FQ216501.

Csnk2a1
Genomic Map