siRNA Details
| Gene Symbol: Ctla2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145799.1 | NP_001139271.1 | 5 | 688 | ||
| NM_007796.2 | NP_031822.2 | 5 | 726 | |||
| GenBank mRNA | AK018801.1 | - | 5 | 681 | ||
| AK132290.1 | - | 5 | 657 | |||
| AK167433.1 | - | 5 | 659 | |||
| AK167664.1 | - | 5 | 660 | |||
| AY034577.1 | - | 5 | 635 | |||
| BC028437.1 | - | 5 | 652 | 1244311 | ||
| X15591.1 | - | 5 | 635 | |||

Ctla2a
Genomic Map