siRNA Details
| Gene Symbol: Ctsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007800.1 | NP_031826.1 | 3 | 397 | ||
| GenBank mRNA | BC125511.1 | - | 3 | 312 | 40130026 | |
| BC125513.1 | - | 3 | 312 | 40130028 | ||
| X78544.1 | - | 3 | 397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsg| Gene Symbol: Ctsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007800.1 | NP_031826.1 | 3 | 397 | ||
| GenBank mRNA | BC125511.1 | - | 3 | 312 | 40130026 | |
| BC125513.1 | - | 3 | 312 | 40130028 | ||
| X78544.1 | - | 3 | 397 | |||