siRNA Details
| Gene Symbol: Ctsk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007802.4 | NP_031828.2 | 2, 3 | 256 | ||
| GenBank mRNA | AK003425.1 | - | 2, 3 | 181 | ||
| AK132648.1 | - | 2, 3 | 186 | |||
| BC046320.1 | - | 2, 3 | 210 | 6390854 | ||
| X94444.1 | - | 1, 2 | 128 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031560, FQ222438, FQ228691, AF010306, BC078793.

Ctsk
Genomic Map