siRNA Details
| Gene Symbol: Ckm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007710.2 | NP_031736.1 | 3 | 338 | ||
| GenBank mRNA | AK009950.1 | - | 3 | 339 | ||
| AK137460.1 | - | 340 | ||||
| BC132424.1 | - | 3 | 336 | 40130701 | ||
| BC132426.1 | - | 3 | 336 | 40130703 | ||
| X03233.1 | - | 3 | 338 | |||

Ckm
Genomic Map