siRNA Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163432.1 | NP_001156904.1 | 3 | 654 | ||
| NM_007712.3 | NP_031738.2 | 3 | 657 | |||
| GenBank mRNA | AF033564.1 | - | 3 | 379 | ||
| AK148377.1 | - | 3 | 644 | |||
| BC015080.1 | - | 3 | 642 | 3995512 | ||
| BY738871.1 | - | 3 | 617 | |||

Clk2
Genomic Map