siRNA Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271607.1 | NP_001258536.1 | 3 | 1440 | ||
| NM_007722.4 | NP_031748.2 | 2 | 1157 | |||
| GenBank mRNA | AF000236.1 | - | 2 | 1111 | ||
| AK031100.1 | - | 1135 | ||||
| BC015254.1 | - | 2 | 1072 | 4242244 | ||

Ackr3
Genomic Map