siRNA Details
| Gene Symbol: Cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007725.2 | NP_031751.1 | 3 | 294 | ||
| GenBank mRNA | AK051845.1 | - | 3 | 295 | ||
| AK218463.1 | - | 2 | 156 | |||
| BC009144.1 | - | 3 | 249 | 3594856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnn2| Gene Symbol: Cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007725.2 | NP_031751.1 | 3 | 294 | ||
| GenBank mRNA | AK051845.1 | - | 3 | 295 | ||
| AK218463.1 | - | 2 | 156 | |||
| BC009144.1 | - | 3 | 249 | 3594856 | ||