siRNA Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020062.3 | NP_064446.2 | 8 | 1704 | ||
| GenBank mRNA | AB044786.1 | - | 6 | 1264 | ||
| AB052777.1 | - | 8 | 1748 | |||
| AF249267.3 | - | 8 | 1649 | |||
| AV659783.2 | - | 1 | 40 | |||
| BC001402.2 | - | 7 | 1475 | |||
| BC017446.1 | - | 5 | 1298 | |||
| BC028349.2 | - | 3 | 899 | |||
| BC052306.1 | - | 8 | 1749 | 6576325 | ||

SLC2A4RG
Genomic Map