siRNA Details
| Gene Symbol: Cplx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007756.3 | NP_031782.3 | 3, 4 | 412 | ||
| GenBank mRNA | AK134820.1 | - | 3, 4 | 355 | ||
| AK158166.1 | - | 3, 4 | 354 | |||
| AK159026.1 | - | 3, 4 | 371 | |||
| BC014803.1 | - | 3, 4 | 317 | 4527117 | ||
| D38614.1 | - | 3, 4 | 344 | |||

Cplx1
Genomic Map