siRNA Details
| Gene Symbol: Crygc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082573.2 | NP_001076042.1 | 3 | 544 | ||
| NM_007775.2 | NP_031801.1 | 3 | 541 | |||
| GenBank mRNA | BC056454.1 | - | 2 | 520 | 6824225 | |
| EF426301.1 | - | 3 | 530 | |||
| EF426302.1 | - | 3 | 533 | |||

Crygc
Genomic Map