siRNA Details
| Gene Symbol: Ctse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007799.3 | NP_031825.2 | 3 | 515 | ||
| GenBank mRNA | AK143581.1 | - | 2 | 217 | ||
| AK145875.1 | - | 3 | 372 | |||
| AK157907.1 | - | 2 | 237 | |||
| AK165242.1 | - | 2 | 237 | |||
| AK165271.1 | - | 2 | 183 | |||
| BC005432.1 | - | 3 | 515 | 3601519 | ||
| CT010252.1 | - | 2 | 151 | |||
| X97399.1 | - | 2 | 202 | |||

Ctse
Genomic Map