siRNA Details
| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289826.1 | NP_001276755.1 | 23 | 3962 | ||
| NM_007715.6 | NP_031741.1 | 23 | 4005 | |||
| GenBank mRNA | AF000998.1 | - | 23 | 4004 | ||
| AK129118.1 | - | 23 | 3962 | |||
| BC129886.1 | - | 21 | 3710 | |||

Clock
Genomic Map