siRNA Details
| Gene Symbol: Ccr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301713.1 | NP_001288642.1 | 2 | 427 | ||
| NM_007719.2 | NP_031745.2 | 3 | 477 | |||
| GenBank mRNA | AK037965.1 | - | 3 | 478 | ||
| AK154349.1 | - | 3 | 478 | |||
| AK154736.1 | - | 3 | 478 | |||
| AK154849.1 | - | 3 | 478 | |||

Ccr7
Genomic Map