siRNA Details
| Gene Symbol: Map3k8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007746.2 | NP_031772.1 | 8 | 1774 | ||
| GenBank mRNA | AK140334.1 | - | 8 | 1775 | ||
| AK149624.1 | - | 8 | 1775 | |||
| AK149846.1 | - | 8 | 1775 | |||
| AK157123.1 | - | 8 | 1741 | |||
| AK171403.1 | - | 7 | 1517 | |||
| AK172424.1 | - | 4 | 908 | |||
| D13759.1 | - | 8 | 1774 | |||

Map3k8
Genomic Map