siRNA Details
| Gene Symbol: Cplx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007756.3 | NP_031782.3 | 4 | 1242 | ||
| GenBank mRNA | AK134820.1 | - | 4 | 1185 | ||
| AK158166.1 | - | 4 | 1185 | |||
| AK159026.1 | - | 4 | 1201 | |||
| BC014803.1 | - | 4 | 1147 | 4527117 | ||

Cplx1
Genomic Map