siRNA Details
| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277231.1 | NP_001264160.1 | 2 | 2702 | ||
| NM_007764.5 | NP_031790.2 | 3 | 3168 | |||
| GenBank mRNA | AK048641.1 | - | 3 | 3166 | ||
| AK087977.1 | - | 1 | 24 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crkl| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277231.1 | NP_001264160.1 | 2 | 2702 | ||
| NM_007764.5 | NP_031790.2 | 3 | 3168 | |||
| GenBank mRNA | AK048641.1 | - | 3 | 3166 | ||
| AK087977.1 | - | 1 | 24 | |||