siRNA Details
| Gene Symbol: ZNF287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020653.2 | NP_065704.2 | 6 | 2926 | ||
| XM_011523968.2 | XP_011522270.1 | 6 | 3372 | |||
| XM_017024887.1 | XP_016880376.1 | 6 | 3133 | |||
| GenBank mRNA | AF217227.1 | - | 6 | 2870 | ||

ZNF287
Genomic Map