siRNA Details
| Gene Symbol: TSHZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136805.1 | - | 1 | 3479 | ||
| BC127095.1 | - | 1 | 3479 | 40123924 | ||
| BC127096.1 | - | 1 | 3479 | 40123925 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSHZ3| Gene Symbol: TSHZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136805.1 | - | 1 | 3479 | ||
| BC127095.1 | - | 1 | 3479 | 40123924 | ||
| BC127096.1 | - | 1 | 3479 | 40123925 | ||