siRNA Details
| Gene Symbol: Cyp3a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007820.2 | NP_031846.2 | 6 | 590 | ||
| GenBank mRNA | BC113143.1 | - | 6 | 555 | 40047455 | |
| D26137.1 | - | 6 | 561 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp3a16| Gene Symbol: Cyp3a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007820.2 | NP_031846.2 | 6 | 590 | ||
| GenBank mRNA | BC113143.1 | - | 6 | 555 | 40047455 | |
| D26137.1 | - | 6 | 561 | |||