siRNA Details
| Gene Symbol: Daxx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199733.1 | NP_001186662.1 | 3 | 920 | ||
| NM_007829.4 | NP_031855.3 | 3 | 820 | |||
| GenBank mRNA | AF006040.1 | - | 2 | 674 | ||
| AK090356.1 | - | 3 | 751 | |||
| AK133863.1 | - | 3 | 728 | |||
| AK144332.1 | - | 3 | 751 | |||
| AK145906.1 | - | 3 | 751 | |||
| AK146743.1 | - | 3 | 747 | |||
| AK151279.1 | - | 3 | 740 | |||
| AK153149.1 | - | 3 | 708 | |||
| AK156425.1 | - | 3 | 921 | |||
| AK172616.1 | - | 3 | 921 | |||
| AY734459.1 | - | 1 | 387 | |||
| AY734461.1 | - | 1 | 304 | |||
| AY734465.1 | - | 1 | 298 | |||
| AY734466.1 | - | 434 | ||||
| AY758273.1 | - | 1 | 302 | |||
| BC128373.1 | - | 3 | 733 | 40111681 | ||
| KJ650207.1 | - | 2 | 650 | |||

Daxx
Genomic Map